Learn More
Apexbio Technology LLC Pizotifen Malate 5189-11-7 300mg
SDP
Catalog No. 5000092987
Encompass_Preferred
Pizotifen malate (CAS 5189-11-7) is a benzocycloheptane derivative that functions as a potent antagonist of serotonin (5-HT) receptors exhibiting high selectivity for this target class By inhibiting serotonergic neurotransmission Pizotifen malate is utilized in biomedical research to investigate the physiological and pharmacological roles of 5-HT pathways Its receptor blocking properties enable studies focused on serotonergic modulation migraine mechanisms and disorders associated with altered serotonin signaling
Catalog No. 50-000-92987 Supplier Apexbio Technology LLC Supplier No. B2007300
Pizotifen malate (CAS 5189-11-7) is a benzocycloheptane derivative that functions as a potent antagonist of serotonin (5-HT) receptors exhibiting high selectivity for this target class By inhibiting serotonergic neurotransmission Pizotifen malate is utilized in biomedical research to investigate the physiological and pharmacological roles of 5-HT pathways Its receptor blocking properties enable studies focused on serotonergic modulation migraine mechanisms and disorders associated with altered serotonin signaling