Learn More
Medchemexpress LLC Pizotifen malate | 5189-11-7 | 99.0% | C23H27NO5S | 100 MG
SDP
Catalog No. 5000432426
Encompass_Preferred
Pizotifen malate (Pizotyline malate) is a potent 5-HT2 receptor antagonist with high affinity for the 5-HT1C binding site. It acts as an antidepressant 5-HT2A receptor antagonist, capable of inhibiting serotonin-enhanced ADP-induced platelet aggregation. This product is intended for research use only and is not sold to patients.
  • Potent 5-HT2 receptor antagonist
  • High affinity for the 5-HT1C binding site
  • Antidepressant 5-HT2A receptor antagonist
  • Inhibits serotonin-enhanced ADP-induced platelet aggregation
  • Intended for research use only
Catalog No. 50-004-32426 Supplier Medchemexpress LLC Supplier No. HYB0115A100MG
Pizotifen malate (Pizotyline malate) is a potent 5-HT2 receptor antagonist with high affinity for the 5-HT1C binding site. It acts as an antidepressant 5-HT2A receptor antagonist, capable of inhibiting serotonin-enhanced ADP-induced platelet aggregation. This product is intended for research use only and is not sold to patients.
  • Potent 5-HT2 receptor antagonist
  • High affinity for the 5-HT1C binding site
  • Antidepressant 5-HT2A receptor antagonist
  • Inhibits serotonin-enhanced ADP-induced platelet aggregation
  • Intended for research use only