Learn More
Description
Includes
13 double-sided pizzas (one side shows fraction) and three double-sided spinners providing different levels of learning.
Seven different games reinforce identifying fractions, matching fraction equivalents, and performing fraction addition and subtraction by building pizzas. For 2-6 players. Grades 1-7.
Order Info
Whole pizzas measure 10 3/4 in..
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School |
| Grade | 1 to 7 |
| For Use With (Application) | Reinforce identifying fractions, matching fraction equivalents, and performing fraction addition and subtraction by building pizzas |
| Includes | 13 double-sided pizzas (one side shows fraction) and three double-sided spinners providing different levels of learning |
| Size | 10.75 in. dia. |
| Diameter (English) Outer | 10.75 in. |
| Class Size | 2 to 6 Students |
| Description | Pizza Fraction Fun Game |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
