Learn More
Enzo Life Sciences PJ-34 (25mg). CAS: 344458-15-7
SDP
Supplier:  Enzo Life Sciences ALX270289M025
Encompass_Preferred
Potent, water soluble poly(ADP-ribose) polymerase (PARP) inhibitor (EC50=20nM compared to EC50=200µM of the prototypical PARP inhibitor 3-aminobenzamide (ALX-270-044)). Inhibits peroxynitrite (ALX-400-036)-induced cell necrosis (EC50=20nM). Has significant, dose-dependent, anti-inflammatory effects in a variety of local inflammation models and provides cardioprotection by decreasing myocardial infarct size. Alternative name: N-(6-Oxo-5,6-dihydro-phenanthridin-2-yl)-N,N-dimethylacetamide . HCl. Formula: C17H17N3O2 . HCl. MW: 295.2 . 36.5. Purity: ≥98% (HPLC). Appearance: White to light brown or yellow powder. Solubility: Soluble in DMSO (up to 30mg/ml) or water (up to 20mg/ml). Long Term Storage: +4°C. Handling: Hygroscopic.
Catalog No. 50-200-8470