Learn More
AdipoGen PJ-34
SDP
Catalog No. 502562366
Encompass_Preferred
Chemical. CAS 344458-15-7. Formula C17H17N3O2 . HCl. MW 295.2 . 36.5. Potent, water soluble polyADP-ribose polymerase PARP inhibitor. Inhibits peroxynitrite-induced cell necrosis. Shows significant, dose-dependent anti-inflammatory effects in a variety of local inflammation models. Provides cardioprotection by decreasing myocardial infarct size. Shows protective effects in models of stroke. Suppresses cell growth in liver cancer cells.
Catalog No. 50-256-2366 Supplier AdipoGen Supplier No. AGCR10100M005
Chemical. CAS 344458-15-7. Formula C17H17N3O2 . HCl. MW 295.2 . 36.5. Potent, water soluble polyADP-ribose polymerase PARP inhibitor. Inhibits peroxynitrite-induced cell necrosis. Shows significant, dose-dependent anti-inflammatory effects in a variety of local inflammation models. Provides cardioprotection by decreasing myocardial infarct size. Shows protective effects in models of stroke. Suppresses cell growth in liver cancer cells.