Learn More
Medchemexpress LLC HY-13688 100mg , PJ34 (hydrochloride) CAS:344458-15-7 Purity:98%
SDP
Catalog No. 501493028
Encompass_Preferred
Medchemexpress, HY-13688 100mg PJ34 (hydrochloride) CAS:344458-15-7 Formula:C17H18ClN3O2 IC50: 110 nM (PARP1), 86 nM (PARP2) Purity:98% PJ34 hydrochloride is a potent specific inhibitor of PARPl / 2 with IC 50 of 110 nM and 86 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3028 Supplier Medchemexpress LLC Supplier No. HY13688100MG
Medchemexpress, HY-13688 100mg PJ34 (hydrochloride) CAS:344458-15-7 Formula:C17H18ClN3O2 IC50: 110 nM (PARP1), 86 nM (PARP2) Purity:98% PJ34 hydrochloride is a potent specific inhibitor of PARPl / 2 with IC 50 of 110 nM and 86 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.