Learn More
Abcam PJ34 hydrochloride, PARP-1 inhibitor, 10MG
Catalog No. 502707066
Encompass_Preferred
MW 331.8 Da, Purity >98%. Potent, selective PARP-1 inhibitor (EC₅₀ = 20 nM). Able to reduce both cell injury induced following deprivation of oxygen-glucose in vitro and infarct size following focal cerebral ischemia in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7066 Supplier Abcam Supplier No. AB12098110MG
MW 331.8 Da, Purity >98%. Potent, selective PARP-1 inhibitor (EC₅₀ = 20 nM). Able to reduce both cell injury induced following deprivation of oxygen-glucose in vitro and infarct size following focal cerebral ischemia in vivo.
The product is subject to the following: Abcam Restricted Use Statement