Learn More
Aviva Systems Biology Corp Pja2 Antibody - N-terminal region (ARP43167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pja2. It was validated on Western Blot. . Alternate names: AI447901, AL022700, MGC27629, Neurodap1, mKIAA0438ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-060