Learn More
Medchemexpress LLC pJAK2(1001-1013) | 1259928-08-9 | 99.9% | 1716.82 | 50 MG
SDP
Catalog No. 5000424944
Encompass_Preferred
pJAK2(1001-1013) is a cell-penetrating peptide that corresponds to the activation loop of JAK2 tyrosine kinase, functioning as a SOCS1/3 antagonist. This peptide blocks SOCS1-mediated negative regulation of immune function and enhances the biological activity of cytokines such as IFNγ and IL6. It has been shown to inhibit the replication of a broad range of viruses and offers dose-dependent protective efficacy against lethal viral infections, making it suitable for studying immune regulation and infection.
  • Functions as a SOCS1/3 antagonist
  • Blocks SOCS1-mediated negative regulation of immune function
  • Enhances biological activity of cytokines like IFNγ and IL6
  • Inhibits replication of a broad range of viruses
  • Offers dose-dependent protective efficacy against lethal viral infections
Catalog No. 50-004-24944 Supplier Medchemexpress LLC Supplier No. HYP1037350MG
Add to Cart
Add to Cart
pJAK2(1001-1013) is a cell-penetrating peptide that corresponds to the activation loop of JAK2 tyrosine kinase, functioning as a SOCS1/3 antagonist. This peptide blocks SOCS1-mediated negative regulation of immune function and enhances the biological activity of cytokines such as IFNγ and IL6. It has been shown to inhibit the replication of a broad range of viruses and offers dose-dependent protective efficacy against lethal viral infections, making it suitable for studying immune regulation and infection.
  • Functions as a SOCS1/3 antagonist
  • Blocks SOCS1-mediated negative regulation of immune function
  • Enhances biological activity of cytokines like IFNγ and IL6
  • Inhibits replication of a broad range of viruses
  • Offers dose-dependent protective efficacy against lethal viral infections