Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pJTI™ R4 EXP CMV-TO EmGFP pA Vector

Catalog No. A15005
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15005 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15005 Supplier LIFE TECHNOLOGIES Supplier No. A15005
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

pJTI™ R4 EXP CMV-TO EmGFP pA Vector

The pJTI™ R4 Exp CMV-TO EmGFP pA vector is a positive control vector for assessing retargeting efficiency when retargeting a Jump-In™ T-REx™ parental cell line. When co-transfected with the integrase vector (pJTI™ R4 Int vector included in the Jump-In™ parental kits) and after antibiotic selection, the EmGFP can be inducibly expressed with doxycycline and the successfully retargeted cells will fluoresce green.

Ensure the Success of Your Jump-In™ T-REx™ Retargeting Reactions
Successful retargeting of Jump-In™ T-REx™ parental cell lines like the Jump-In™ T-REx™ HEK293 Kit is dependent on a variety of factors, such as:

  • Transfection efficiency
  • Cell confluency
  • Antibiotic selection conditions
  • Quality and concentration of DNA
  • Retargeting vector to integrase vector ratio

    We strongly recommend including a positive control retargeting reaction using the pJTI™ R4 Exp CMV-TO EmGFP pA vector in your Jump-In™ experiment, along with negative controls (no plasmid DNA, no integrase vector), so you can easily visualize the results and optimize the retargeting conditions.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Constitutive or Inducible System Inducible
    Delivery Type Transfection
    Inducing Agent Tetracycline
    Promoter CMV/TO
    Product Type Mammalian Expression Vector
    Reporter Gene GFP (EmGFP)
    Selection Agent (Eukaryotic) Geneticin™ (G-418)
    Content And Storage pJTI™ R4 EXP CMV-TO EmGFP pA vector (100 μg), store at -20 °C
    Antibiotic Resistance Bacterial Ampicillin (AmpR)
    Protein Tag GFP (EmGFP)
    Quantity 100 μg
    Vector Jump-In Vectors
    Product Line Jump-In
    For Use With (Application) Stable Cell Line Development, Targeted Integration, Regulated Expression
    Show More Show Less
    WARNING: Reproductive Harm - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.