Learn More
Enzo Life Sciences pJY2 plasmid (5µg)
SDP
Catalog No. 502010521
Encompass_Preferred
The AGA arginine codon is rarely used in E. coli but is common in eukaryotic genes. The low level of tRNAUCUArg can lead to low expression and misincorporation of lysine for arginine, during expression of genes containing AGA codons in E. coli. E. coli harboring pJY2 can be employed to suppress lysine misincorporation and achieve high-level expression of genes cloned into pETvectors without modification of AGA codons in the target gene sequence. pJY2 encodes tRNAUCUArg (to suppress lysine misincorporation at AGA codons) and T7 lysozyme (to depress constitutive expression of the cloned gene). For example, pJY2 allows for faithful expression of pET3a-encoded mutant ubiquitins with a high density of AGA codons (up to 14% of total codons). pJY2 can also be used as a source of tRNAUCUArg with expression systems, such as the pGEX series of vectors, that do not rely on T7 polymerase. Formulation: Liquid. In 10mM TRIS, pH 8.0, containing 1mM EDTA. Long Term Storage: -20°C.
Catalog No. 50-201-0521 Supplier Enzo Life Sciences Supplier No. BMLDW87200005
The AGA arginine codon is rarely used in E. coli but is common in eukaryotic genes. The low level of tRNAUCUArg can lead to low expression and misincorporation of lysine for arginine, during expression of genes containing AGA codons in E. coli. E. coli harboring pJY2 can be employed to suppress lysine misincorporation and achieve high-level expression of genes cloned into pETvectors without modification of AGA codons in the target gene sequence. pJY2 encodes tRNAUCUArg (to suppress lysine misincorporation at AGA codons) and T7 lysozyme (to depress constitutive expression of the cloned gene). For example, pJY2 allows for faithful expression of pET3a-encoded mutant ubiquitins with a high density of AGA codons (up to 14% of total codons). pJY2 can also be used as a source of tRNAUCUArg with expression systems, such as the pGEX series of vectors, that do not rely on T7 polymerase. Formulation: Liquid. In 10mM TRIS, pH 8.0, containing 1mM EDTA. Long Term Storage: -20°C.