Learn More
Metal Working Chemical & Equipment Co Easicult Combi dip slide culture test kit, dual-sided, for microbial contamination monitoring, 10 tests per box
Catalog No. NC9678808
Encompass
Easicult Combi dip slide culture test kit, dual-sided, for microbial contamination monitoring, 10 tests per box
Catalog No. NC9678808 Supplier Metal Working Chemical & Equipment Co Supplier No. 4060
Easicult Combi dip slide culture test kit, dual-sided, for microbial contamination monitoring, 10 tests per box