Learn More
Enzo Life Sciences PK-11195 (5mg). CAS: 85532-75-8
SDP
Supplier:  Enzo Life Sciences BMLCM1180005
Encompass_Preferred
PK-11195 is a ligand of the mitochondrial benzodiazepine receptor and selective antagonist of benzodiazepine. It facilitates the induction of apoptosis and reverses Bcl-2-mediated cytoprotection. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, methanol, dichlormethane or DMSO. Insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0422