Learn More
AdipoGen PK 11195
SDP
Catalog No. 502562399
Encompass_Preferred
Chemical. CAS 85532-75-8. Formula C21H21ClN2O. MW 352.9. Selective peripheral benzodiazepine antagonist. Apoptosis enhancer. Glucose-induced insulin secretion inhibitor. Induces mitochondria cytochrome c release. Anticancer compound. Antiproliferative. Pharmacological tool in autophagy.
Catalog No. 50-256-2399 Supplier AdipoGen Supplier No. AGCR10008M010
Chemical. CAS 85532-75-8. Formula C21H21ClN2O. MW 352.9. Selective peripheral benzodiazepine antagonist. Apoptosis enhancer. Glucose-induced insulin secretion inhibitor. Induces mitochondria cytochrome c release. Anticancer compound. Antiproliferative. Pharmacological tool in autophagy.