Learn More
Andwin Scientific PK/12 64-CELL DIVIDERS (8X8)
SDP
Catalog No. NC9567335
Encompass_Preferred
The 64-cell dividers perform as a cell partitions for carboard storage boxes. This product measures 8 x 8.
Catalog No. NC9567335 Supplier Andwin Scientific Supplier No. H1276
The 64-cell dividers perform as a cell partitions for carboard storage boxes. This product measures 8 x 8.