Learn More
IDEX Health & Science Upchurch PK ADPT 1/4 ML NPT - 1/4-28FM
Catalog No. NC1920542
Encompass_Preferred
Idex National Pipe Thread (NPT) Adapters, For Use With: 0.25 in. Male NPT to 0.25-28 Female, Includes: P-300/308 for 1/8 in. OD tubing, PEEK, NPT Adapter, Red, Diameter Inner: 0.062 in.
Catalog No. NC1920542 Supplier IDEX Health & Science Upchurch Supplier No. U500
Idex National Pipe Thread (NPT) Adapters, For Use With: 0.25 in. Male NPT to 0.25-28 Female, Includes: P-300/308 for 1/8 in. OD tubing, PEEK, NPT Adapter, Red, Diameter Inner: 0.062 in.