Learn More
Ace Glass, Inc. 60/50 ground joint standard taper PTFE sleeve
SDP
Catalog No. 50975203
Encompass
60/50 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 1.
Catalog No. 50-975-203 Supplier Ace Glass, Inc. Supplier No. 764225
60/50 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 1.