Learn More
Ace Glass, Inc. PK1 71/60 PTFE SLEEVE PK1 71/60 PTFE SLEEVE
SDP
Catalog No. 50979389
Encompass
71/60 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005¿). Pack of 1.
Catalog No. 50-979-389 Supplier Ace Glass, Inc. Supplier No. 764227
71/60 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005¿). Pack of 1.