Learn More
Medchemexpress LLC HY-U00447 5mg Medchemexpress, PK11007 CAS:38275-34-2 Purity:>98%
SDP
Catalog No. 501934455
Encompass_Preferred
Medchemexpress, HY-U00447 5mg PK11007 CAS:38275-34-2 PK11007 is a p53 targeting compound, has anti-tumor activities through activation of unstable p53 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4455 Supplier Medchemexpress LLC Supplier No. HYU004475MG
Medchemexpress, HY-U00447 5mg PK11007 CAS:38275-34-2 PK11007 is a p53 targeting compound, has anti-tumor activities through activation of unstable p53 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.