Learn More
Medchemexpress LLC PK11007 | 874146-69-7 | 98.7% | 10 MG
SDP
Catalog No. 5000342938
Encompass_Preferred
PK11007 is a mild thiol alkylator with anticancer activity. It stabilizes p53 through selective alkylation of two surface-exposed cysteines without affecting its DNA binding activity. PK11007 also induces cell death in mutant p53 cancer cells by increasing reactive oxygen species (ROS) levels.
  • Reduces viability significantly in mutant p53 cell lines
  • Induces mainly caspase-independent cell death
  • Up-regulates protein levels of p53 target genes (p21, MDM2, PUMA)
  • Increases p53 activity
  • Increases transcription of p53 target genes
  • Elevates ROS levels
  • Inhibits cell proliferation, induces apoptosis, and alters genes involved in cell death
Catalog No. 50-003-42938 Supplier Medchemexpress LLC Supplier No. HY12878410MG
PK11007 is a mild thiol alkylator with anticancer activity. It stabilizes p53 through selective alkylation of two surface-exposed cysteines without affecting its DNA binding activity. PK11007 also induces cell death in mutant p53 cancer cells by increasing reactive oxygen species (ROS) levels.
  • Reduces viability significantly in mutant p53 cell lines
  • Induces mainly caspase-independent cell death
  • Up-regulates protein levels of p53 target genes (p21, MDM2, PUMA)
  • Increases p53 activity
  • Increases transcription of p53 target genes
  • Elevates ROS levels
  • Inhibits cell proliferation, induces apoptosis, and alters genes involved in cell death