Learn More
Medchemexpress LLC PK11007 | 874146-69-7 | 98.7% | 50 MG
SDP
Catalog No. 5000342868
Encompass_Preferred
PK11007 is a mild thiol alkylator with anticancer activity. It stabilizes p53 and induces cell death in mutant p53 cancer cells by elevating reactive oxygen species (ROS) levels. It inhibits cell proliferation, induces apoptosis, and alters genes involved in cell death, consistent with its ability to reactivate mutant p53.
  • Mild thiol alkylator
  • Exhibits anticancer activity
  • Stabilizes p53
  • Induces cell death in mutant p53 cancer cells
  • Elevates reactive oxygen species (ROS) levels
  • Inhibits cell proliferation
  • Induces apoptosis
  • Alters genes involved in cell death
  • Reactivates mutant p53
Catalog No. 50-003-42868 Supplier Medchemexpress LLC Supplier No. HY12878450MG
Add to Cart
Add to Cart
PK11007 is a mild thiol alkylator with anticancer activity. It stabilizes p53 and induces cell death in mutant p53 cancer cells by elevating reactive oxygen species (ROS) levels. It inhibits cell proliferation, induces apoptosis, and alters genes involved in cell death, consistent with its ability to reactivate mutant p53.
  • Mild thiol alkylator
  • Exhibits anticancer activity
  • Stabilizes p53
  • Induces cell death in mutant p53 cancer cells
  • Elevates reactive oxygen species (ROS) levels
  • Inhibits cell proliferation
  • Induces apoptosis
  • Alters genes involved in cell death
  • Reactivates mutant p53