Learn More
AdipoGen PK 11195 (50 mg)
SDP
Supplier:  AdipoGen AGCR10008M050
Encompass_Preferred
Chemical. CAS: 85532-75-8. Formula: C21H21ClN2O. MW: 352.9. Selective peripheral benzodiazepine antagonist. Apoptosis enhancer. Glucose-induced insulin secretion inhibitor. Induces mitochondria cytochrome c release. Anticancer compound. Antiproliferative. Pharmacological tool in autophagy.
Catalog No. 50-205-3756