Learn More
Abcam PK11195, benzodiazepine receptor antagonist, 10MG
Catalog No. 502707635
Encompass_Preferred
MW 352.9 Da, Purity >99%. Antagonist at the mitochondrial (peripheral) benzodiazepine receptor (translocator protein, TSPO). Acts as a chemosensitizer of tumor cells to a wide range of chemotherapeutic agents.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7635 Supplier Abcam Supplier No. AB12037810MG
MW 352.9 Da, Purity >99%. Antagonist at the mitochondrial (peripheral) benzodiazepine receptor (translocator protein, TSPO). Acts as a chemosensitizer of tumor cells to a wide range of chemotherapeutic agents.
The product is subject to the following: Abcam Restricted Use Statement