Learn More
ATCC PK(15); Kidney; Pig (Sus scrofa)
Supplier:  ATCC CCL33
Encompass
PK(15) [PK15, PK-15] is a cell line exhibiting epithelial morphology that was isolated from the kidney of an adult pig.
Catalog No. 50-238-2460