Learn More
Ace Glass, Inc. PK2 2-WAY VALVE TEFZEL PK2 2-WAY VALVE TEFZEL
SDP
Catalog No. 50983434
Encompass
Omnifit two-way tubing connector valve with single Tefzel control valve, rated at 500 psi. Valve has one moving part for long life and trouble-free operation. Unique design prevents the valve from leaking to outside, even at excessive pressures. Body is machined from a single piece of PTFE. Caps are made of polypropylene with Fluoron elastomer seals for instant connection of rigid or flexible tubing within a range of 0.5 to 4mm O.D. Connector has a ¿¿-28 thread at the back of the cap for direct connection to other adapters. Autoclaveable. Pack of 2.
Catalog No. 50-983-434 Supplier Ace Glass, Inc. Supplier No. 1272832
Omnifit two-way tubing connector valve with single Tefzel control valve, rated at 500 psi. Valve has one moving part for long life and trouble-free operation. Unique design prevents the valve from leaking to outside, even at excessive pressures. Body is machined from a single piece of PTFE. Caps are made of polypropylene with Fluoron elastomer seals for instant connection of rigid or flexible tubing within a range of 0.5 to 4mm O.D. Connector has a ¿¿-28 thread at the back of the cap for direct connection to other adapters. Autoclaveable. Pack of 2.