Learn More
Ace Glass, Inc. 10/10 ground joint standard taper PTFE sleeve
SDP
Catalog No. 50975200
Encompass
10/10 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 3.
Catalog No. 50-975-200 Supplier Ace Glass, Inc. Supplier No. 764204
10/10 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 3.