Learn More
Ace Glass, Inc. PK3 14/23 PTFE SLEEVE PK3 14/23 PTFE SLEEVE
SDP
Catalog No. 50975204
Encompass
14/23 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005¿). Pack of 3.
Catalog No. 50-975-204 Supplier Ace Glass, Inc. Supplier No. 7642114
14/23 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005¿). Pack of 3.