Learn More
Ace Glass, Inc. 24/29 ground joint standard taper PTFE sleeve
SDP
Catalog No. 50975205
Encompass
24/29 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 3.
Catalog No. 50-975-205 Supplier Ace Glass, Inc. Supplier No. 7642124
24/29 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 3.