Learn More
Ace Glass, Inc. 45/40 ground joint standard taper PTFE sleeve, Pack of 3
SDP
Catalog No. 50975206
Encompass
45/40 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 3.
Catalog No. 50-975-206 Supplier Ace Glass, Inc. Supplier No. 7642145
45/40 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005). Pack of 3.