Learn More
TARGETMOL CHEMICALS INC PK68 5MG
SDP
Catalog No. 502581448
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PK68 is an effective and selective inhibitor of type II receptor-interacting kinase 1 (RIPK1 IC50 = 90 nM). PK68 can be used for research on the treatment of inflammatory disorders and cancer metastasis. purity: 98%
Catalog No. 50-258-1448 Supplier TARGETMOL CHEMICALS INC Supplier No. T124935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PK68 is an effective and selective inhibitor of type II receptor-interacting kinase 1 (RIPK1 IC50 = 90 nM). PK68 can be used for research on the treatment of inflammatory disorders and cancer metastasis. purity: 98%