Learn More
Enzo Life Sciences PKA alpha (catalytic domain) (mouse), (recombinant) (5000 U)
SDP
Supplier:  Enzo Life Sciences BMLSE1925000
Encompass_Preferred
Alternative name: cAMP dependent protein kinase alpha, Protein kinase A alpha. Concentration: ~0.5mg/ml. Formulation: Liquid. In 50% glycerol buffer. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-1954