Learn More
Enzo Life Sciences PKA kinase activity kit (1x96wells)
SDP
Catalog No. 50659121
Encompass_Preferred
Non-radioactive protein kinase assay provides rapid and sensitive detection. The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio. Alternative Name: Protein kinase A. cAMP-dependent protein kinase. Applications: Colorimetric detection, Activity assay. Species Reactivity: Species independent. Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2
Catalog No. 50-659-121 Supplier Enzo Life Sciences Supplier No. ADIEKS390A
Non-radioactive protein kinase assay provides rapid and sensitive detection. The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio. Alternative Name: Protein kinase A. cAMP-dependent protein kinase. Applications: Colorimetric detection, Activity assay. Species Reactivity: Species independent. Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2