Learn More
Enzo Life Sciences PKA (CT) polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKASPK017D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human PKA. UniProt ID: P17612 (Cα subunit), P22694 (Cβ subunit), P22162 (Cγ subunit), P10644 (RIα subunit), P31321 (RIβ subunit), P13861 (RIIα subunit), P31323 (RIIβ subunit). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7319