Learn More
Medchemexpress LLC HY-P1290A 5mg Medchemexpress, PKA Inhibitor Fragment (6-22) amide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1290A5MG
Encompass_Preferred
Medchemexpress, HY-P1290A 5mg PKA Inhibitor Fragment (6-22) amide (TFA) CAS: PKA Inhibitor Fragment (6-22) amide TFA is an inhibitor of cAMP-dependent protein kinase A (PKA), with a Ki of 2.8 nM. PKA Inhibitor Fragment (6-22) amide TFA can significantly reverse low-level morphine antinociceptive tolerance in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3048