Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PKA RII peptide TFA | 99.7% | 2226.37 | 10 MG
SDP

Catalog No. 5000425366
Encompass_Preferred

PKA RII peptide TFA is a PKA substrate that, after being phosphorylated at the serine residue, can be used for the detection of calcineurin activity. It is for research use only.

  • Solid appearance, white to off-white color.
  • Sequence: Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-R-Val-Ser-Val-Ala-Ala-Glu.
  • Sequence shortening: DLDVPIPGRFDRRVSVAAE.
  • Store as powder at -80°C for 2 years or -20°C for 1 year.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).
  • Soluble in H2O at 25 mg/mL (11.23 mM, requires ultrasonic).

Catalog No. 50-004-25366 Supplier Medchemexpress LLC Supplier No. HYP0228A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PKA RII peptide TFA is a PKA substrate that, after being phosphorylated at the serine residue, can be used for the detection of calcineurin activity. It is for research use only.

  • Solid appearance, white to off-white color.
  • Sequence: Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-R-Val-Ser-Val-Ala-Ala-Glu.
  • Sequence shortening: DLDVPIPGRFDRRVSVAAE.
  • Store as powder at -80°C for 2 years or -20°C for 1 year.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).
  • Soluble in H2O at 25 mg/mL (11.23 mM, requires ultrasonic).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.