Learn More
Carnabio Usa Inc PKACA(PRKACA)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 011275
Encompass
PKAC(PRKACA), Full-length, Wild type, Amino Acid 1-351, NP_002721.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0579