Learn More
Eurofins Discoverx Akt1/PKBα (δPH, S473D), active, 10 µg. (14-453)
Supplier:  Eurofins Discoverx 14453
Encompass
N-terminal 6xHis-tagged recombinant, human Akt1, amino acids 118-end containing the mutation S473D. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8817