Learn More
Eurofins Discoverx Akt2/PKBβ (δPH, S474D), active, 10 µg. (14-339)
Supplier:  Eurofins Discoverx 14339
Encompass
N-terminal 6xHis-tagged recombinant, human PKBβ, amino acids 120-481, S474D; activated by PDK1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8775