Learn More
Eurofins Discoverx Akt3/PKBγ (S472D), active, 250 µg. (14-502M)
Supplier:  Eurofins Discoverx 14502M
Encompass
N-terminal 6xHis-tagged recombinant, human Akt3/PKBγ amino acids 117-end, containing a S472D mutation. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8878