Learn More
Enzo Life Sciences PKC alpha (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4940005
Encompass_Preferred
Alternative name: Protein kinase C alpha. Source: Produced in insect cells. Full length active PKCa (protein kinase C α) is fused at the N-terminus to a GST-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2052