Learn More
Eurofins Discoverx PKCε, active, 10 µg. (14-518)
Supplier:  Eurofins Discoverx 14518
Encompass
N-terminal 6xHis-tagged recombinant PKCε amino acids 2-end. PKC Lipid activator (Catalog # 20-133) is required for use. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8905