Learn More
Eurofins Discoverx PKC ζ, active, 10 µg. (14-525)
Supplier:  Eurofins Discoverx 14525
Encompass
N-terminal 6xHis-tagged recombinant, human PKC zeta, amino acids 2-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8919