Learn More
Enzo Life Sciences PKC-alpha monoclonal antibody (MC5) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMPK020F
Encompass_Preferred
Clone: MC5. Host: Mouse. Isotype: IgG2a. Immunogen: Bovine brain PKCα. UniProt ID: P04409. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Gerbil, Fish, Dog, Bovine. Applications: Flow Cytometry, IF, IHC, IP, WB. Recommended Dilutions: Flow Cytometry (10µg/ml) Immunohistochemistry (1:50, human lung, carcinoma tissue) Immunoprecipitation (12.5µg/ml) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7256