Learn More
Enzo Life Sciences PKC alpha polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA1440100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 662-672 of rabbit PKCα (protein kinase C α). Species reactivity: Human, Mouse, Rat Mammalian. Applications: WB. Formulation: Neat serum. Contains 0.05% sodium azide.
Catalog No. 50-201-1762