Learn More
Sino Biological Recombinant Human PDK2 Protein (Full Length, GST Tag), Active
Catalog No. 502573597
Encompass_Preferred
Recombinant full length human PDHK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3597 Supplier Sino Biological Supplier No. P5810G50
Recombinant full length human PDHK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.