Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PKCα (C2-4) inhibitor peptide | 99.5% | 1107.17 | 10 MG
SDP

Catalog No. 5000399846
Encompass_Preferred

The MedChemExpress HY-P5881 is a PKCα (C2-4) inhibitor peptide, appearing as a white to off-white solid. It has a molecular weight of 1107.17 and demonstrates a high purity of 99.5% via LCMS analysis. This product is intended for research applications, requiring specific storage conditions to maintain its stability.

  • Specific PKCα inhibitor peptide
  • White to off-white solid appearance
  • Molecular weight: 1107.17
  • Purity (LCMS): 99.5%
  • For research use only

Catalog No. 50-003-99846 Supplier Medchemexpress LLC Supplier No. HYP588110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MedChemExpress HY-P5881 is a PKCα (C2-4) inhibitor peptide, appearing as a white to off-white solid. It has a molecular weight of 1107.17 and demonstrates a high purity of 99.5% via LCMS analysis. This product is intended for research applications, requiring specific storage conditions to maintain its stability.

  • Specific PKCα inhibitor peptide
  • White to off-white solid appearance
  • Molecular weight: 1107.17
  • Purity (LCMS): 99.5%
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.