Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PKCα (C2-4) inhibitor peptide | 99.5% | 1107.17 | 25 MG
SDP

Catalog No. 5000397423
Encompass_Preferred

PKCα (C2-4) inhibitor peptide is a synthetic peptide that blocks protein kinase C (PKC) activity. This peptide is suitable for various research applications requiring specific inhibition of PKC activity.

  • Effectively blocks protein kinase C activity in HepG2 cells when stimulated with calcium and pancreastatin at 10 μM.
  • Purity is 99.5%.
  • Appearance is a white to off-white solid.
  • Soluble in DMF (1 mg/ml), DMSO (10 mg/ml), and PBS (pH 7.2, 5 mg/ml).
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year.
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).

Catalog No. 50-003-97423 Supplier Medchemexpress LLC Supplier No. 25MGHYP5881
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PKCα (C2-4) inhibitor peptide is a synthetic peptide that blocks protein kinase C (PKC) activity. This peptide is suitable for various research applications requiring specific inhibition of PKC activity.

  • Effectively blocks protein kinase C activity in HepG2 cells when stimulated with calcium and pancreastatin at 10 μM.
  • Purity is 99.5%.
  • Appearance is a white to off-white solid.
  • Soluble in DMF (1 mg/ml), DMSO (10 mg/ml), and PBS (pH 7.2, 5 mg/ml).
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year.
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.