Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pkcα (c2-4) inhibitor peptide | 99.5% | 1107.17 | 50 MG
SDP

Catalog No. 5000397906
Encompass_Preferred

The PKCα (C2-4) inhibitor peptide, identified by MedChemExpress with catalog number HY-P5881, is a high-purity chemical compound for research use. It is a peptide that specifically inhibits PKCα (C2-4). The product has a purity of 99.5% confirmed by LCMS analysis and a molecular weight of 1107.17.

  • White to off-white solid appearance
  • High purity of 99.5% (LCMS)
  • Molecular weight of 1107.17
  • Stable for 2 years at -80°C as powder
  • Stable for 1 year at -20°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent

Catalog No. 50-003-97906 Supplier Medchemexpress LLC Supplier No. HYP588150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PKCα (C2-4) inhibitor peptide, identified by MedChemExpress with catalog number HY-P5881, is a high-purity chemical compound for research use. It is a peptide that specifically inhibits PKCα (C2-4). The product has a purity of 99.5% confirmed by LCMS analysis and a molecular weight of 1107.17.

  • White to off-white solid appearance
  • High purity of 99.5% (LCMS)
  • Molecular weight of 1107.17
  • Stable for 2 years at -80°C as powder
  • Stable for 1 year at -20°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.