Learn More
Enzo Life Sciences PKC delta substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1540001
Encompass_Preferred
This peptide sequence, ARRKRKGSFFYGG, was selected from a peptide library for optimal phosphorylation by protein kinase C δ (PKCδ). Alternative name: Protein kinase C delta substrate. Purity: ≥95% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1262