Learn More
Enzo Life Sciences [Ser159] PKC epsilon (149-164) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP1550500
Encompass_Preferred
Protein kinase C ε (PKCε) pseudosubstrate sequence (149-164) but with an Ala to Ser159 substitution (ERMRPRKRQGSVRRRV) which makes it a substrate for phosphorylation. Phospholipid/phorbol ester-dependent substrate for PKC ε, ζ, δ and η. Alternative name: [Ser159] Protein kinase C epsilon (149 164), PKC epsilon substrate. Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1263